2,3-Difluoro-6-nitrotoluene 95%

BD162606-25G BD162606-250MG BD162606-1G BD162606-5G BD162606-10G BD162606-100MG
BLDPBD162606-25GEA 527 EUR
BLDPBD162606-25G BLDPBD162606-250MG BLDPBD162606-1G BLDPBD162606-5G BLDPBD162606-10G BLDPBD162606-100MG
2,3-Difluoro-6-nitrotoluene 95%
2,3-Difluoro-6-nitrotoluene

Storage condition: Sealed in dry, Room Temperature

Formel: C₇H₅F₂NO₂
Molecular Weight: 173,117 g/mol
Lagertemperatur: Ambient
MDL: MFCD08706407
CAS-Nummer: 914348-35-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR