5-Hydroxyisophthalamide 98%

BD162571-1G BD162571-250MG BD162571-5G
BLDPBD162571-1GEA 58 EUR
BLDPBD162571-1G BLDPBD162571-250MG BLDPBD162571-5G
5-Hydroxyisophthalamide 98%
5-Hydroxyisophthalamide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₈H₈N₂O₃
Molecular Weight: 180,16 g/mol
Lagertemperatur: Ambient
MDL: MFCD16038667
CAS-Nummer: 68052-43-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR