3-Amino-4-chlor-1H-indazol 97%

BLDPBD162522-5GEA 84 EUR
BLDPBD162522-5G BLDPBD162522-25G BLDPBD162522-250MG BLDPBD162522-1G
3-Amino-4-chlor-1H-indazol 97%
3-Amino-4-chloro-1H-indazole

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₇H₆ClN₃
Molecular Weight: 167,60 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00663139
CAS-Nummer: 20925-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR