2-Acetylphenanthrene 95%

BD16252-100MG BD16252-250MG BD16252-5G BD16252-1G
BLDPBD16252-100MGEA 19 EUR
BLDPBD16252-100MG BLDPBD16252-250MG BLDPBD16252-5G BLDPBD16252-1G
2-Acetylphenanthrene 95%
2-Acetylphenanthrene

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₂O
Molecular Weight: 220,2700 g/mol
Lagertemperatur: Ambient
MDL: MFCD00001171
CAS-Nummer: 5960-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR