5-Fluoro-2-methylpyrimidine-4,6-diol 96%

Synonyme: 5-Fluoro-6-hydroxy-2-methyl-4(1H)-pyrimidinone

BD162501-100MG BD162501-250MG
BLDPBD162501-100MGEA 17 EUR
BLDPBD162501-100MG BLDPBD162501-250MG
5-Fluoro-2-methylpyrimidine-4,6-diol 96%
5-Fluoro-2-methylpyrimidine-4,6-diol

Storage condition: Sealed in dry, 2-8°C

Formel: C₅H₅FN₂O₂
Molecular Weight: 144,11 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11520459
CAS-Nummer: 1598-63-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR