(S)-Methyl-2,3-dihydroxypropanoate 97%

Synonyme: (S)-Methyl 2,3-dihydroxypropanoate

BD162467-25G BD162467-5G BD162467-1G BD162467-250MG BD162467-10G
BLDPBD162467-25GEA 534 EUR
BLDPBD162467-25G BLDPBD162467-5G BLDPBD162467-1G BLDPBD162467-250MG BLDPBD162467-10G
(S)-Methyl-2,3-dihydroxypropanoate 97%
(S)-Methyl-2,3-dihydroxypropanoate

Storage condition: Sealed in dry, Store in freezer, under -20°C

Formel: C₄H₈O₄
Molecular Weight: 120,1039 g/mol
Lagertemperatur: Freezer
MDL: MFCD14636488
CAS-Nummer: 10303-88-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR