2-Chloro-4-cyanophenylboronic acid pinacol ester 97%

Synonyme: 3-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

BD162409-250MG BD162409-1G
BLDPBD162409-250MGEA 74 EUR
BLDPBD162409-250MG BLDPBD162409-1G
2-Chloro-4-cyanophenylboronic acid pinacol ester 97%
2-Chloro-4-cyanophenylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₅BClNO₂
Molecular Weight: 263,528 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD18072641
CAS-Nummer: 945391-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR