4-(3,4-Dichlorophenyl)-1-tetralone 97%

Synonyme: 4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one

BD16240-10G BD16240-25G BD16240-100G BD16240-5G
BLDPBD16240-10GEA 9 EUR
BLDPBD16240-10G BLDPBD16240-25G BLDPBD16240-100G BLDPBD16240-5G
4-(3,4-Dichlorophenyl)-1-tetralone 97%
4-(3,4-Dichlorophenyl)-1-tetralone

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₂Cl₂O
Molecular Weight: 291,18 g/mol
Lagertemperatur: Ambient
MDL: MFCD02093088
CAS-Nummer: 79560-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR