N,N-Dibenzyl-4-bromoaniline 95%

Synonyme: 4-Bromo-N,N-dibenzylaniline

BD162329-250MG BD162329-1G BD162329-5G
BLDPBD162329-250MGEA 31 EUR
BLDPBD162329-250MG BLDPBD162329-1G BLDPBD162329-5G
N,N-Dibenzyl-4-bromoaniline 95%
N,N-Dibenzyl-4-bromoaniline

Storage condition: Inert atmosphere, Room Temperature

Formel: C₂₀H₁₈BrN
Molecular Weight: 352,28 g/mol
Lagertemperatur: Ambient
MDL: MFCD09037473
CAS-Nummer: 65145-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR