Methyl-3-(4-bromphenyl)propanoat 98%

Synonyme: Methyl 3-(4-Bromophenyl)propionate,3-(4-Bromophenyl)propionic acid methyll ester,Methyl 3-(4-bromophenyl)propanoate

BD162322-100G BD162322-10G BD162322-5G BD162322-25G BD162322-1G
BLDPBD162322-100GEA 302 EUR
BLDPBD162322-100G BLDPBD162322-10G BLDPBD162322-5G BLDPBD162322-25G BLDPBD162322-1G
Methyl-3-(4-bromphenyl)propanoat 98%
Methyl-3-(4-bromophenyl)propanoate

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₀H₁₁BrO₂
Molecular Weight: 243,10 g/mol
Lagertemperatur: Ambient
MDL: MFCD09953150
CAS-Nummer: 75567-84-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR