4-(Pyridin-2-yl)butan-1-ol 97%

BD162278-100MG BD162278-5G BD162278-10G BD162278-250MG BD162278-1G
BLDPBD162278-100MGEA 54 EUR
BLDPBD162278-100MG BLDPBD162278-5G BLDPBD162278-10G BLDPBD162278-250MG BLDPBD162278-1G
4-(Pyridin-2-yl)butan-1-ol 97%
4-(Pyridin-2-yl)butan-1-ol

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₃NO
Molecular Weight: 151,206 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD12828284
CAS-Nummer: 17945-79-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR