[1,1'-Biphenyl]-3,3',4,4'-tetracarboxylic acid 97%

BD16222-5G BD16222-250MG BD16222-25G BD16222-1G
BLDPBD16222-5GEA 117 EUR
BLDPBD16222-5G BLDPBD16222-250MG BLDPBD16222-25G BLDPBD16222-1G
[1,1'-Biphenyl]-3,3',4,4'-tetracarboxylic acid 97%
[1,1'-Biphenyl]-3,3',4,4'-tetracarboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₀O₈
Molecular Weight: 330,25 g/mol
Lagertemperatur: Ambient
MDL: MFCD00496632
CAS-Nummer: 22803-05-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR