2-(7-Methoxynaphthalen-1-yl)ethanamine hydrochloride 97%

BD162215-5G BD162215-25G BD162215-10G BD162215-250MG BD162215-1G
BLDPBD162215-5GEA 79 EUR
BLDPBD162215-5G BLDPBD162215-25G BLDPBD162215-10G BLDPBD162215-250MG BLDPBD162215-1G
2-(7-Methoxynaphthalen-1-yl)ethanamine hydrochloride 97%
2-(7-Methoxynaphthalen-1-yl)ethanamine hydrochloride

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₃H₁₆ClNO
Molecular Weight: 237,72 g/mol
Lagertemperatur: Ambient
MDL: MFCD13185884
CAS-Nummer: 139525-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR