Benzo[b]naphtho[1,2-d]furan 97%

Synonyme: Naphtho[2,1-b]benzofuran

BD16220-5G BD16220-250MG BD16220-25G BD16220-100MG BD16220-1G
BLDPBD16220-5GEA 214 EUR
BLDPBD16220-5G BLDPBD16220-250MG BLDPBD16220-25G BLDPBD16220-100MG BLDPBD16220-1G
Benzo[b]naphtho[1,2-d]furan 97%
Benzo[b]naphtho[1,2-d]furan

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₀O
Molecular Weight: 218,26 g/mol
Lagertemperatur: Ambient
MDL: MFCD00046916
CAS-Nummer: 205-39-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR