1-Ethyl-1H-pyrazole-5-carbaldehyde 95+%

BLDPBD162196-1GEA 65 EUR
BLDPBD162196-1G BLDPBD162196-5G BLDPBD162196-250MG
1-Ethyl-1H-pyrazole-5-carbaldehyde 95+%
1-Ethyl-1H-pyrazole-5-carbaldehyde

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₈N₂O
Molecular Weight: 124,14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD03130036
CAS-Nummer: 902837-62-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR