(5-Bromopyrimidin-2-yl)methanamine 98+%

BD162135-100MG BD162135-250MG BD162135-1G
BLDPBD162135-100MGEA 176 EUR
BLDPBD162135-100MG BLDPBD162135-250MG BLDPBD162135-1G
(5-Bromopyrimidin-2-yl)methanamine 98+%
(5-Bromopyrimidin-2-yl)methanamine
Formel: C₅H₆BrN₃
Molecular Weight: 188,03 g/mol
MDL: MFCD15071122
CAS-Nummer: 1240783-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR