1'-Methylspiro[indoline-3,4'-piperidine] 95%

BLDPBD162119-1GEA 529 EUR
BLDPBD162119-1G BLDPBD162119-100MG BLDPBD162119-250MG
1'-Methylspiro[indoline-3,4'-piperidine] 95%
1'-Methylspiro[indoline-3,4'-piperidine]

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formel: C₁₃H₁₈N₂
Molecular Weight: 202,2954 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12198567
CAS-Nummer: 69584-91-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR