2-(4-Bromophenyl)quinoline-4-carboxylic acid 95+%

BD16209-10G BD16209-5G BD16209-250MG BD16209-1G
BLDPBD16209-10GEA 824 EUR
BLDPBD16209-10G BLDPBD16209-5G BLDPBD16209-250MG BLDPBD16209-1G
2-(4-Bromophenyl)quinoline-4-carboxylic acid 95+%
2-(4-Bromophenyl)quinoline-4-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₆H₁₀BrNO₂
Lagertemperatur: Ambient
MDL: MFCD00160560
CAS-Nummer: 103914-52-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR