2-Bromo-1,1-dimethoxypropane 95%

BD162084-100G BD162084-1G BD162084-250MG BD162084-10G BD162084-5G BD162084-25G
BLDPBD162084-100GEA 1104 EUR
BLDPBD162084-100G BLDPBD162084-1G BLDPBD162084-250MG BLDPBD162084-10G BLDPBD162084-5G BLDPBD162084-25G
2-Bromo-1,1-dimethoxypropane 95%
2-Bromo-1,1-dimethoxypropane

Storage condition: Inert atmosphere, Room Temperature

Formel: C₅H₁₁BrO₂
Lagertemperatur: Ambient
MDL: MFCD11505877
CAS-Nummer: 33170-72-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR