1-(Methoxymethyl)-1H-imidazole 98%

BD162043-250MG
BLDPBD162043-250MGEA 8 EUR
BLDPBD162043-250MG
1-(Methoxymethyl)-1H-imidazole 98%
1-(Methoxymethyl)-1H-imidazole

Storage condition: Sealed in dry, Room Temperature

Formel: C₅H₈N₂O
Molecular Weight: 112,12 g/mol
Lagertemperatur: Ambient
MDL: MFCD00955700
CAS-Nummer: 20075-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR