1H-Pyrazol-1-ol 97%

BD162032-1G BD162032-25G BD162032-5G BD162032-10G
BLDPBD162032-1GEA 46 EUR
BLDPBD162032-1G BLDPBD162032-25G BLDPBD162032-5G BLDPBD162032-10G
1H-Pyrazol-1-ol 97%
1H-Pyrazol-1-ol

Storage condition: Sealed in dry, Room Temperature

Formel: C₃H₄N₂O
Molecular Weight: 84,08 g/mol
Lagertemperatur: Ambient
MDL: MFCD11850516
CAS-Nummer: 81945-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR