Valdecoxib 99+%

Synonyme: 4-(5-methyl-3-phenylisoxazol-4-yl)benzenesulphonamide

BD16203-5G BD16203-1G BD16203-25G BD16203-100G BD16203-250MG
BLDPBD16203-5GEA 66 EUR
BLDPBD16203-5G BLDPBD16203-1G BLDPBD16203-25G BLDPBD16203-100G BLDPBD16203-250MG
Valdecoxib 99+%
Valdecoxib

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₆H₁₄N₂O₃S
Molecular Weight: 314,36 g/mol
Lagertemperatur: Ambient
MDL: MFCD00950568
CAS-Nummer: 181695-72-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR