9-Phenanthrencarbonitril 98%

Synonyme: 9-Cyanphenanthren

BD16202-1G BD16202-5G BD16202-25G BD16202-250MG
BLDPBD16202-1GEA 55 EUR
BLDPBD16202-1G BLDPBD16202-5G BLDPBD16202-25G BLDPBD16202-250MG
9-Phenanthrencarbonitril 98%
9-Phenanthrencarbonitril

Storage condition: Sealed in dry, Room Temperature

Formel: C₁₅H₉N
Molecular Weight: 203,24 g/mol
Lagertemperatur: Ambient
MDL: MFCD00001173
CAS-Nummer: 2510-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR