N-(5-Chloro-2-hydroxyphenyl)acetamide 97%

BD162018-250MG BD162018-100MG
BLDPBD162018-250MGEA 174 EUR
BLDPBD162018-250MG BLDPBD162018-100MG
N-(5-Chloro-2-hydroxyphenyl)acetamide 97%
N-(5-Chloro-2-hydroxyphenyl)acetamide

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₈ClNO₂
Molecular Weight: 185,60 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00029752
CAS-Nummer: 26488-93-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR