1-Bromo-3-methylbutan-2-one 95%

BD162016-100MG BD162016-250MG BD162016-1G BD162016-5G BD162016-10G
BLDPBD162016-100MGEA 13 EUR
BLDPBD162016-100MG BLDPBD162016-250MG BLDPBD162016-1G BLDPBD162016-5G BLDPBD162016-10G
1-Bromo-3-methylbutan-2-one 95%
1-Bromo-3-methylbutan-2-one

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₅H₉BrO
Molecular Weight: 165,0284 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD00015716
CAS-Nummer: 19967-55-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR