9-Anthracenylmethyl acrylate 98+%

Synonyme: Anthracen-9-ylmethyl acrylate,Acrylic Acid 9-Anthrylmethyl Ester,Anthracen-9-ylmethyl Prop-2-enoate,2-propenoic acid 9-anthracenylmethyl ester,9-Anthrylmethyl Acrylate

BD162012-100MG BD162012-25G BD162012-5G BD162012-1G BD162012-250MG
BLDPBD162012-100MGEA 26 EUR
BLDPBD162012-100MG BLDPBD162012-25G BLDPBD162012-5G BLDPBD162012-1G BLDPBD162012-250MG
9-Anthracenylmethyl acrylate 98+%
9-Anthracenylmethyl acrylate

Storage condition: Inert atmosphere, Room Temperature

Formel: C₁₈H₁₄O₂
Molecular Weight: 262,30 g/mol
Lagertemperatur: Ambient
MDL: MFCD04039960
CAS-Nummer: 31645-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR