5-Bromo-2-chlorobenzothiazole 98%

Synonyme: 5-Bromo-2-chlorobenzo[d]thiazole

BD161953-5G BD161953-250MG BD161953-1G
BLDPBD161953-5GEA 319 EUR
BLDPBD161953-5G BLDPBD161953-250MG BLDPBD161953-1G
5-Bromo-2-chlorobenzothiazole 98%
5-Bromo-2-chlorobenzothiazole

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₃BrClNS
Molecular Weight: 248,53 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09749227
CAS-Nummer: 824403-26-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR