(4-Ethoxy-2-formylphenyl)boronic acid 95+%

BLDPBD161837-5GEA 267 EUR
BLDPBD161837-5G BLDPBD161837-250MG BLDPBD161837-1G
(4-Ethoxy-2-formylphenyl)boronic acid 95+%
(4-Ethoxy-2-formylphenyl)boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₉H₁₁BO₄
Molecular Weight: 193,99 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08458481
CAS-Nummer: 1106867-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR