tert-Butyl 3-acetylpyrrolidine-1-carboxylate 98%

BD161785-1G BD161785-100MG BD161785-250MG BD161785-5G
BLDPBD161785-1GEA 177 EUR
BLDPBD161785-1G BLDPBD161785-100MG BLDPBD161785-250MG BLDPBD161785-5G
tert-Butyl 3-acetylpyrrolidine-1-carboxylate 98%
tert-Butyl 3-acetylpyrrolidine-1-carboxylate

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₁H₁₉NO₃
Molecular Weight: 213,27 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD11865235
CAS-Nummer: 858643-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR