3,5-Difluoro-4-(hydroxymethyl)benzoic acid 95%

BD161784-10G BD161784-250MG BD161784-100MG BD161784-5G BD161784-1G
BLDPBD161784-10GEA 1085 EUR
BLDPBD161784-10G BLDPBD161784-250MG BLDPBD161784-100MG BLDPBD161784-5G BLDPBD161784-1G
3,5-Difluoro-4-(hydroxymethyl)benzoic acid 95%
3,5-Difluoro-4-(hydroxymethyl)benzoic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆F₂O₃
Molecular Weight: 188,12 g/mol
Lagertemperatur: Ambient
MDL: MFCD12547172
CAS-Nummer: 1211596-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR