(4-Bromo-3-methylphenyl)acetic acid 98%

Synonyme: 2-(4-Bromo-3-methylphenyl)acetic acid

BD161763-5G BD161763-25G BD161763-100G BD161763-10G BD161763-250MG BD161763-1G
BLDPBD161763-5GEA 113 EUR
BLDPBD161763-5G BLDPBD161763-25G BLDPBD161763-100G BLDPBD161763-10G BLDPBD161763-250MG BLDPBD161763-1G
(4-Bromo-3-methylphenyl)acetic acid 98%
(4-Bromo-3-methylphenyl)acetic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₉BrO₂
Molecular Weight: 229,08 g/mol
Lagertemperatur: Ambient
MDL: MFCD02664811
CAS-Nummer: 215949-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR