1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene 95%

BLDPBD161757-1GEA 29 EUR
BLDPBD161757-1G
1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene 95%
1,3-Difluoro-4-nitro-2-(trifluoromethyl)benzene

Storage condition: Sealed in dry, 2-8°C

Formel: C₇H₂F₅NO₂
Molecular Weight: 227,09 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12924861
CAS-Nummer: 123973-36-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR