5-Bromo-1-chloro-6-methylisoquinoline 96%

BD161701-1G BD161701-5G BD161701-100MG BD161701-250MG
BLDPBD161701-1GEA 285 EUR
BLDPBD161701-1G BLDPBD161701-5G BLDPBD161701-100MG BLDPBD161701-250MG
5-Bromo-1-chloro-6-methylisoquinoline 96%
5-Bromo-1-chloro-6-methylisoquinoline

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₀H₇BrClN
Molecular Weight: 256,52 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09264576
CAS-Nummer: 1245647-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR