7-Fluoroquinolin-8-ol 98%

BD161681-100MG
BLDPBD161681-100MGEA 41 EUR
BLDPBD161681-100MG
7-Fluoroquinolin-8-ol 98%
7-Fluoroquinolin-8-ol
Formel: C₉H₆FNO
Molecular Weight: 163,148 g/mol
MDL: MFCD12024552
CAS-Nummer: 35048-10-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR