2-Amino-4-methyl-5-pyrimidinboronsäurepinakolester 95%

Synonyme: 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine,4-Methyl-2-aminopyrimidine-5-boronic acid, pinacol ester

BD161633-1G BD161633-100MG BD161633-250MG
BLDPBD161633-1GEA 341 EUR
BLDPBD161633-1G BLDPBD161633-100MG BLDPBD161633-250MG
2-Amino-4-methyl-5-pyrimidinboronsäurepinakolester 95%
2-Amino-4-methyl-5-pyrimidinboronsäurepinakolester

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Formel: C₁₁H₁₈BN₃O₂
Molecular Weight: 235,09 g/mol
Lagertemperatur: Freezer
MDL: MFCD18072554
CAS-Nummer: 944401-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR