3-Benzyloxy-6-brom-2-fluorphenylboronsäure 97%

Synonyme: 3-(Benzyloxy)-6-bromo-2-fluorophenylboronic acid

BD161605-1G BD161605-25G BD161605-10G BD161605-5G
BLDPBD161605-1GEA 15 EUR
BLDPBD161605-1G BLDPBD161605-25G BLDPBD161605-10G BLDPBD161605-5G
3-Benzyloxy-6-brom-2-fluorphenylboronsäure 97%
3-Benzyloxy-6-brom-2-fluorphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₁BBrFO₃
Molecular Weight: 324,9400 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD09475868
CAS-Nummer: 957035-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR