5-Carboxy-2-fluorphenylboronsäure 96%

Synonyme: 3-Borono-4-fluorobenzoic acid

BD161581-5G BD161581-10G BD161581-25G BD161581-250MG BD161581-1G
BLDPBD161581-5GEA 140 EUR
BLDPBD161581-5G BLDPBD161581-10G BLDPBD161581-25G BLDPBD161581-250MG BLDPBD161581-1G
5-Carboxy-2-fluorphenylboronsäure 96%
5-Carboxy-2-fluorphenylboronsäure

Storage condition: Inert atmosphere, Room Temperature

Formel: C₇H₆BFO₄
Molecular Weight: 183,9300 g/mol
Lagertemperatur: Ambient
MDL: MFCD08436065
CAS-Nummer: 874219-59-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR