3-Benzyloxy-5-fluorphenylboronsäure 97%

Synonyme: 3-Benzyloxy-5-fluorobenzeneboronic acid

BD161572-250MG BD161572-1G
BLDPBD161572-250MGEA 29 EUR
BLDPBD161572-250MG BLDPBD161572-1G
3-Benzyloxy-5-fluorphenylboronsäure 97%
3-Benzyloxy-5-fluorphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₃H₁₂BFO₃
Molecular Weight: 246,04 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD07363782
CAS-Nummer: 850589-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR