5-Isopropyl-2-methyl-2H-pyrazole-3-carboxylic acid 98%

Synonyme: 3-Isopropyl-1-methyl-1H-pyrazole-5-carboxylic acid

BD161561-250MG BD161561-5G BD161561-1G
BLDPBD161561-250MGEA 27 EUR
BLDPBD161561-250MG BLDPBD161561-5G BLDPBD161561-1G
5-Isopropyl-2-methyl-2H-pyrazole-3-carboxylic acid 98%
5-Isopropyl-2-methyl-2H-pyrazole-3-carboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formel: C₈H₁₂N₂O₂
Molecular Weight: 168,2 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD07186363
CAS-Nummer: 78208-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR