2-Chloro-5-(trifluoromethyl)pyridine-3-boronic acid 98%

BD161548-250MG BD161548-100MG BD161548-1G
BLDPBD161548-250MGEA 24 EUR
BLDPBD161548-250MG BLDPBD161548-100MG BLDPBD161548-1G
2-Chloro-5-(trifluoromethyl)pyridine-3-boronic acid 98%
2-Chloro-5-(trifluoromethyl)pyridine-3-boronic acid

Storage condition: Inert atmosphere, 2-8°C

Formel: C₆H₄BClF₃NO₂
Molecular Weight: 225,36 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD10696241
CAS-Nummer: 536693-96-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR