3-Formyl-2-methoxyphenylboronsäure 95%

Lieferant: BLD PHARMATECH GMBH

BLDPBD161542-1GEA 23 EUR
BLDPBD161542-1G BLDPBD161542-100MG BLDPBD161542-250MG
3-Formyl-2-methoxyphenylboronsäure 95%
3-Formyl-2-methoxyphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₈H₉BO₄
Molecular Weight: 179,9700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08689486
CAS-Nummer: 480424-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR