2,6-Difluor-4-methoxyphenylboronsäure 97%

BLDPBD161540-5GEA 58 EUR
BLDPBD161540-5G BLDPBD161540-10G BLDPBD161540-1G BLDPBD161540-100G BLDPBD161540-25G BLDPBD161540-250MG
2,6-Difluor-4-methoxyphenylboronsäure 97%
2,6-Difluor-4-methoxyphenylboronsäure

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formel: C₇H₇BF₂O₃
Molecular Weight: 187,94 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05664299
CAS-Nummer: 406482-20-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR