2-Chlor-6-fluor-3-methylphenylboronsäure 95%

BD161537-1G BD161537-250MG BD161537-5G
BLDPBD161537-1GEA 18 EUR
BLDPBD161537-1G BLDPBD161537-250MG BLDPBD161537-5G
2-Chlor-6-fluor-3-methylphenylboronsäure 95%
2-Chlor-6-fluor-3-methylphenylboronsäure

Storage condition: Inert atmosphere, 2-8°C

Formel: C₇H₇BClFO₂
Molecular Weight: 188,3900 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD03701534
CAS-Nummer: 352535-85-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR