(S)-tert-Butyl-1-methyl-2-oxoimidazolidine-4-carboxylate 97%

Synonyme: (S)-tert-Butyl 1-methyl-2-oxoimidazolidine-4-carboxylate

BD161490-10G BD161490-25G BD161490-5G BD161490-250MG BD161490-1G
BLDPBD161490-10GEA 74 EUR
BLDPBD161490-10G BLDPBD161490-25G BLDPBD161490-5G BLDPBD161490-250MG BLDPBD161490-1G
(S)-tert-Butyl-1-methyl-2-oxoimidazolidine-4-carboxylate 97%
(S)-tert-Butyl-1-methyl-2-oxoimidazolidine-4-carboxylate

Storage condition: Sealed in dry, Room Temperature

Formel: C₉H₁₆N₂O₃
Molecular Weight: 200,24 g/mol
Lagertemperatur: Ambient
MDL: MFCD09952527
CAS-Nummer: 83056-79-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR