Isorhapontigenin 95%

Synonyme: (E)-5-(4-hydroxy-3-methoxystyryl)benzene-1,3-diol,3,4',5-Trihydroxy-3'-methoxy-trans-stilbene

BD161470-100MG
BLDPBD161470-100MGEA 509 EUR
BLDPBD161470-100MG
Isorhapontigenin 95%
Isorhapontigenin

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formel: C₁₅H₁₄O₄
Molecular Weight: 258,27 g/mol
Lagertemperatur: Refrigerator
MDL: MFCD12407151
CAS-Nummer: 32507-66-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR