Piceatannol 98%

Lieferant: BLD PHARMATECH GMBH

BLDPBD161462-250MGEA 26 EUR
BLDPBD161462-250MG BLDPBD161462-100MG BLDPBD161462-5G BLDPBD161462-10G BLDPBD161462-25G BLDPBD161462-1G
Piceatannol 98%
Piceatannol

Storage condition: Sealed in dry, 2-8°C

Formel: C₁₄H₁₂O₄
Molecular Weight: 244,2427 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00221715
CAS-Nummer: 10083-24-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR