Xylazole hydrochloride 98%

BLDPBD161432-250MGEA 137 EUR
BLDPBD161432-250MG BLDPBD161432-1G BLDPBD161432-100MG
Xylazole hydrochloride 98%
Xylazole hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formel: C₁₁H₁₃ClN₂S
Molecular Weight: 240,75 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD13184267
CAS-Nummer: 123941-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR