2-Amino-N-(2,2,2-trifluoroethyl)acetamide 95%

BLDPBD161428-100MGEA 6 EUR
BLDPBD161428-100MG BLDPBD161428-25G BLDPBD161428-250MG BLDPBD161428-5G BLDPBD161428-1G
2-Amino-N-(2,2,2-trifluoroethyl)acetamide 95%
2-Amino-N-(2,2,2-trifluoroethyl)acetamide

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₄H₇F₃N₂O
Molecular Weight: 156,106 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09944516
CAS-Nummer: 359821-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR