2-Aminobenzo[d]thiazole-7-carboxylic acid 97%

BD161369-100MG BD161369-250MG
BLDPBD161369-100MGEA 69 EUR
BLDPBD161369-100MG BLDPBD161369-250MG
2-Aminobenzo[d]thiazole-7-carboxylic acid 97%
2-Aminobenzo[d]thiazole-7-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formel: C₈H₆N₂O₂S
Molecular Weight: 194,2104 g/mol
Lagertemperatur: Ambient
MDL: MFCD13195418
CAS-Nummer: 71224-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR