2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine 95%

BLDPBD161335-100MGEA 65 EUR
BLDPBD161335-100MG BLDPBD161335-250MG BLDPBD161335-5G BLDPBD161335-1G
2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine 95%
2-(4-Methylpiperazin-1-yl)pyrimidin-5-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formel: C₉H₁₅N₅
Molecular Weight: 193,2489 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13193411
CAS-Nummer: 943757-74-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR